Learn More
Meso Scale Discovery VPLEX Plus MSE IL27p28/IL30
SDP
Catalog No. 5000081867
Encompass
The Meso Scale Discovery IL-27p28/IL-30 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-27p28 and IL-30 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results with a broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to complete the assay The kit can quantify IL-27p28 and IL-30 simultaneously in the same sample in under 5 hours
Catalog No. 50-000-81867 Supplier Meso Scale Discovery Supplier No. K152WXG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery IL-27p28/IL-30 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-27p28 and IL-30 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results with a broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to complete the assay The kit can quantify IL-27p28 and IL-30 simultaneously in the same sample in under 5 hours