Learn More
Meso Scale Discovery VPLEX Plus MSE IL27p28/IL30
SDP
Catalog No. 5000081865
Encompass
The Meso Scale Discovery V-PLEX Mouse Cytokine 30-Plex Kit is an electrochemiluminescent immunoassay for measuring 30 mouse cytokines chemokines and growth factors in various biological fluids It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators controls diluent wash buffer read buffer and plate seals The assay features a dynamic range of 3-4 logs high sensitivity and requires 25-50 L of sample It runs on MSD s SECTOR Imager for efficient cytokine profiling
Catalog No. 50-000-81865 Supplier Meso Scale Discovery Supplier No. K152WXG1
The Meso Scale Discovery V-PLEX Mouse Cytokine 30-Plex Kit is an electrochemiluminescent immunoassay for measuring 30 mouse cytokines chemokines and growth factors in various biological fluids It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators controls diluent wash buffer read buffer and plate seals The assay features a dynamic range of 3-4 logs high sensitivity and requires 25-50 L of sample It runs on MSD s SECTOR Imager for efficient cytokine profiling