Learn More
Meso Scale Discovery VPLEX Plus NHP Cytokine 24Plex
SDP
Catalog No. 5000078311
Encompass
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a sensitive immunoassay for measuring human interleukin 6 (IL-6) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for accurate IL-6 detection at sub-picogram levels The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Customizable components optimize it for specific samples and research With high sensitivity and minimal interference it is ideal for inflammation and immune response studies
Catalog No. 50-000-78311 Supplier Meso Scale Discovery Supplier No. K15058G2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a sensitive immunoassay for measuring human interleukin 6 (IL-6) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for accurate IL-6 detection at sub-picogram levels The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Customizable components optimize it for specific samples and research With high sensitivity and minimal interference it is ideal for inflammation and immune response studies