Learn More
Meso Scale Discovery VPLEX Plus NHP Cytokine 24Plex
SDP
Supplier:  Meso Scale Discovery K15058G1
Encompass
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a highly sensitive and quantitative assay for measuring human interleukin 6 (IL-6) in serum plasma and cell culture supernatants This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to enable simultaneous detection of IL-6 down to 0 04 pg/mL with an upper limit of 1500 pg/mL The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium calibrators diluent wash buffer read buffer and adhesive plate seals sufficient for one 96-well plate This assay provides accurate reproducible quantitation of IL-6 in just 3 5 hours
Catalog No. 50-000-81695