Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 1 NHP Kit
SDP
Catalog No. 5000078264
Encompass
The Meso Scale Discovery V-PLEX Plus Viral Panel 1 NHP Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum plasma and lung lavage samples This assay kit includes a 96-well plate detection antibodies wash buffers calibrators controls and other reagents needed to run the immunoassay on the MSD platform The V-PLEX Plus Viral Panel 1 NHP Kit allows for high-throughput quantitative antibody profiling of NHP samples enabling researchers to efficiently monitor immune responses to respiratory viruses in preclinical studies
Catalog No. 50-000-78264 Supplier Meso Scale Discovery Supplier No. K15348G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Viral Panel 1 NHP Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum plasma and lung lavage samples This assay kit includes a 96-well plate detection antibodies wash buffers calibrators controls and other reagents needed to run the immunoassay on the MSD platform The V-PLEX Plus Viral Panel 1 NHP Kit allows for high-throughput quantitative antibody profiling of NHP samples enabling researchers to efficiently monitor immune responses to respiratory viruses in preclinical studies