Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 1 NHP Kit
SDP
Catalog No. 5000081509
Encompass
The Meso Scale Discovery V-PLEX Plus Viral Panel 1 NHP Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum plasma and lung lavage samples This kit includes a 96-well plate coated with antigens for the viruses detection antibodies calibrators controls diluent wash buffer read buffer and plate seals The electrochemiluminescence immunoassay allows for high sensitivity and a broad dynamic range This kit can help researchers studying immune responses to respiratory viruses in non-human primate models
Catalog No. 50-000-81509 Supplier Meso Scale Discovery Supplier No. K15348G1
The Meso Scale Discovery V-PLEX Plus Viral Panel 1 NHP Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum plasma and lung lavage samples This kit includes a 96-well plate coated with antigens for the viruses detection antibodies calibrators controls diluent wash buffer read buffer and plate seals The electrochemiluminescence immunoassay allows for high sensitivity and a broad dynamic range This kit can help researchers studying immune responses to respiratory viruses in non-human primate models