Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 1 NHP Kit
SDP
Catalog No. 5000082136
Encompass
The Meso Scale Diagnostics V-PLEX Plus Viral Panel 1 NHP Kit is a multiplex immunoassay panel that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum plasma and lung lavage samples The kit includes a 96-well plate coated with antigens detection antibodies calibrators controls diluent wash buffer read buffer and plate seals The electrochemiluminescence immunoassay allows for high sensitivity and a broad dynamic range This kit is for research use only and not for use in diagnostic procedures It provides a fast and efficient way to profile viral antibody responses in non-human primate studies
Catalog No. 50-000-82136 Supplier Meso Scale Discovery Supplier No. K15348G2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Diagnostics V-PLEX Plus Viral Panel 1 NHP Kit is a multiplex immunoassay panel that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum plasma and lung lavage samples The kit includes a 96-well plate coated with antigens detection antibodies calibrators controls diluent wash buffer read buffer and plate seals The electrochemiluminescence immunoassay allows for high sensitivity and a broad dynamic range This kit is for research use only and not for use in diagnostic procedures It provides a fast and efficient way to profile viral antibody responses in non-human primate studies