Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 1HMNKit
SDP
Catalog No. 5000080352
Encompass
The Meso Scale Discovery V-PLEX Plus Viral Panel 1 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in human serum and plasma This assay kit includes a 96-well plate coated with antigens for the viruses detection antibodies assay diluent wash buffer read buffer and calibrators to quantitatively determine antibody titers The electrochemiluminescent technology allows high sensitivity and a broad dynamic range This product enables efficient serological testing for current or past exposure to common respiratory viruses
Catalog No. 50-000-80352 Supplier Meso Scale Discovery Supplier No. K15345G1
The Meso Scale Discovery V-PLEX Plus Viral Panel 1 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in human serum and plasma This assay kit includes a 96-well plate coated with antigens for the viruses detection antibodies assay diluent wash buffer read buffer and calibrators to quantitatively determine antibody titers The electrochemiluminescent technology allows high sensitivity and a broad dynamic range This product enables efficient serological testing for current or past exposure to common respiratory viruses