Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 1HMNKit
SDP
Catalog No. 5000082050
Encompass
Meso Scale Discovery s V-PLEX Plus Cytokine Panel 1 (human) kit measures 10 cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 TNF- ) from various samples This electrochemiluminescence immunoassay uses patented MULTI-ARRAY technology with V-PLEX Plus plates and an optional QuickPlex SQ 120 instrument for fast analysis The kit includes all reagents offers high sensitivity a wide dynamic range and requires low sample volume providing a customizable multiplex immunoassay for biomarker research
Catalog No. 50-000-82050 Supplier Meso Scale Discovery Supplier No. K15345G2
Add to Cart
Add to Cart
Request a Quote
Meso Scale Discovery s V-PLEX Plus Cytokine Panel 1 (human) kit measures 10 cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 TNF- ) from various samples This electrochemiluminescence immunoassay uses patented MULTI-ARRAY technology with V-PLEX Plus plates and an optional QuickPlex SQ 120 instrument for fast analysis The kit includes all reagents offers high sensitivity a wide dynamic range and requires low sample volume providing a customizable multiplex immunoassay for biomarker research