Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 1HMNKit
SDP
Catalog No. 5000082051
Encompass
The Meso Scale Discovery V-PLEX Plus Viral Panel 1 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in human serum and plasma This assay uses electrochemiluminescence detection technology and provides quantitative results for each analyte in under 3 hours The kit includes a 96-well plate coated with antigens detection antibodies calibrators controls wash buffer read buffer and plate seals This assay can aid in the diagnosis of viral respiratory infections
Catalog No. 50-000-82051 Supplier Meso Scale Discovery Supplier No. K15345G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Viral Panel 1 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in human serum and plasma This assay uses electrochemiluminescence detection technology and provides quantitative results for each analyte in under 3 hours The kit includes a 96-well plate coated with antigens detection antibodies calibrators controls wash buffer read buffer and plate seals This assay can aid in the diagnosis of viral respiratory infections