Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 2 NHP Kit
SDP
Catalog No. 5000078272
Encompass
The Meso Scale Discovery V-PLEX Plus Viral Panel 2 (NHP) Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum and plasma samples This assay utilizes electrochemiluminescence detection technology on patterned arrays for high sensitivity and specificity The kit includes a 96-well plate detection antibodies wash buffers calibrators controls and other reagents needed to run the assay It can provide quantitative results in under 5 hours to aid in viral disease diagnosis and research This kit is for research use only and not for use in diagnostic procedures
Catalog No. 50-000-78272 Supplier Meso Scale Discovery Supplier No. K15349G1
The Meso Scale Discovery V-PLEX Plus Viral Panel 2 (NHP) Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum and plasma samples This assay utilizes electrochemiluminescence detection technology on patterned arrays for high sensitivity and specificity The kit includes a 96-well plate detection antibodies wash buffers calibrators controls and other reagents needed to run the assay It can provide quantitative results in under 5 hours to aid in viral disease diagnosis and research This kit is for research use only and not for use in diagnostic procedures