Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 2 NHP Kit
SDP
Catalog No. 5000079378
Encompass
The Meso Scale Discovery V-PLEX Non-Human Primate IFN-gamma Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IFN-gamma in non-human primate serum plasma and culture supernatants The assay utilizes MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of up to 3 logs and shows minimal cross-reactivity The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrators and controls Sample volume is 25 L The assay can be read on MSD s SECTOR Imager and results calculated with MSD Discovery Workbench software
Catalog No. 50-000-79378 Supplier Meso Scale Discovery Supplier No. K15349G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Non-Human Primate IFN-gamma Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IFN-gamma in non-human primate serum plasma and culture supernatants The assay utilizes MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of up to 3 logs and shows minimal cross-reactivity The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrators and controls Sample volume is 25 L The assay can be read on MSD s SECTOR Imager and results calculated with MSD Discovery Workbench software