Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 2 NHP Kit
SDP
Catalog No. 5000082052
Encompass
The V-PLEX Plus Viral Panel 2 (NHP) Kit from Meso Scale Diagnostics is a multiplex immunoassay kit that allows for the simultaneous detection of antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum plasma and lung lavage samples The kit contains a 96-well plate coated with antigens for the viruses detection antibodies assay diluent wash buffer read buffer and a plate seal The electrochemiluminescence immunoassay allows for high sensitivity and a broad dynamic range This kit can aid in viral surveillance and vaccine studies utilizing non-human primate models
Catalog No. 50-000-82052 Supplier Meso Scale Discovery Supplier No. K15349G2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Viral Panel 2 (NHP) Kit from Meso Scale Diagnostics is a multiplex immunoassay kit that allows for the simultaneous detection of antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum plasma and lung lavage samples The kit contains a 96-well plate coated with antigens for the viruses detection antibodies assay diluent wash buffer read buffer and a plate seal The electrochemiluminescence immunoassay allows for high sensitivity and a broad dynamic range This kit can aid in viral surveillance and vaccine studies utilizing non-human primate models