Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 2HMNKit
SDP
Catalog No. 5000079376
Encompass
The Meso Scale Discovery V-PLEX Plus Viral Panel 2 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in human serum and plasma This assay kit contains a 96-well plate coated with antigens for the viruses detection antibodies buffers calibrators controls and other reagents needed to run the immunoassay The electrochemiluminescence technology allows for high sensitivity and a broad dynamic range This product can aid in diagnosis of viral respiratory infections and is for research use only
Catalog No. 50-000-79376 Supplier Meso Scale Discovery Supplier No. K15346G1
The Meso Scale Discovery V-PLEX Plus Viral Panel 2 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in human serum and plasma This assay kit contains a 96-well plate coated with antigens for the viruses detection antibodies buffers calibrators controls and other reagents needed to run the immunoassay The electrochemiluminescence technology allows for high sensitivity and a broad dynamic range This product can aid in diagnosis of viral respiratory infections and is for research use only