Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 2HMNKit
SDP
Catalog No. 5000079806
Encompass
The Meso Scale Discovery V-PLEX Plus Viral Panel 2 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in human serum and plasma This assay kit contains a 96-well plate coated with antigens for the viruses detection antibodies standards controls diluent wash buffer read buffer and plate seals The electrochemiluminescence technology allows sensitive and accurate quantification of antibody levels This product enables efficient serological testing for current or past exposure to common respiratory viruses
Catalog No. 50-000-79806 Supplier Meso Scale Discovery Supplier No. K15346G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Viral Panel 2 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in human serum and plasma This assay kit contains a 96-well plate coated with antigens for the viruses detection antibodies standards controls diluent wash buffer read buffer and plate seals The electrochemiluminescence technology allows sensitive and accurate quantification of antibody levels This product enables efficient serological testing for current or past exposure to common respiratory viruses