Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 3 NHP Kit
SDP
Catalog No. 5000082054
Encompass
The Meso Scale Discovery V-PLEX Plus Viral Panel 3 NHP Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Virus and Influenza B Virus in non-human primate serum and plasma samples This assay utilizes electrochemiluminescence detection technology and provides quantitative results for each analyte in a single well The kit includes a 96-well plate coated with antigens detection antibodies calibrators controls wash buffer read buffer and plate seals sufficient for testing 90 samples in duplicate This assay can aid in the differential diagnosis of respiratory infections in non-human primate studies
Catalog No. 50-000-82054 Supplier Meso Scale Discovery Supplier No. K15350G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Viral Panel 3 NHP Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Virus and Influenza B Virus in non-human primate serum and plasma samples This assay utilizes electrochemiluminescence detection technology and provides quantitative results for each analyte in a single well The kit includes a 96-well plate coated with antigens detection antibodies calibrators controls wash buffer read buffer and plate seals sufficient for testing 90 samples in duplicate This assay can aid in the differential diagnosis of respiratory infections in non-human primate studies