Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 3 NHP Kit
SDP
Catalog No. 5000080380
Encompass
The V-PLEX Viral Panel 3 (NHP) Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for the simultaneous detection of antibodies to SARS-CoV-2 Influenza A Virus and Influenza B Virus in non-human primate serum and plasma samples The kit contains a 96-well plate coated with antigens for the viruses detection antibodies assay diluent wash buffer read buffer and calibrators and controls The electrochemiluminescence immunoassay allows high sensitivity and a broad dynamic range This kit allows efficient screening of non-human primate samples for evidence of exposure to these viruses
Catalog No. 50-000-80380 Supplier Meso Scale Discovery Supplier No. K15350G2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Viral Panel 3 (NHP) Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for the simultaneous detection of antibodies to SARS-CoV-2 Influenza A Virus and Influenza B Virus in non-human primate serum and plasma samples The kit contains a 96-well plate coated with antigens for the viruses detection antibodies assay diluent wash buffer read buffer and calibrators and controls The electrochemiluminescence immunoassay allows high sensitivity and a broad dynamic range This kit allows efficient screening of non-human primate samples for evidence of exposure to these viruses