Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 3 NHP Kit
SDP
Catalog No. 5000082143
Encompass
The V-PLEX Plus Viral Panel 3 NHP Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for the simultaneous detection of antibodies to SARS-CoV-2 Influenza A Virus and Influenza B Virus in non-human primate serum and plasma samples The kit contains a 96-well plate coated with viral antigens detection antibodies calibrators controls diluent wash buffer read buffer and plate seals The electrochemiluminescent assay allows high sensitivity and a broad dynamic range This kit is for research use only and not for use in diagnostic procedures It allows efficient profiling of antibody responses to multiple viruses in non-human primate studies
Catalog No. 50-000-82143 Supplier Meso Scale Discovery Supplier No. K15350G1
The V-PLEX Plus Viral Panel 3 NHP Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for the simultaneous detection of antibodies to SARS-CoV-2 Influenza A Virus and Influenza B Virus in non-human primate serum and plasma samples The kit contains a 96-well plate coated with viral antigens detection antibodies calibrators controls diluent wash buffer read buffer and plate seals The electrochemiluminescent assay allows high sensitivity and a broad dynamic range This kit is for research use only and not for use in diagnostic procedures It allows efficient profiling of antibody responses to multiple viruses in non-human primate studies