Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 3HMNKit
SDP
Catalog No. 5000078267
Encompass
The Meso Scale Discovery V-PLEX Plus Viral Panel 3 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A and Influenza B in human serum and plasma This assay uses electrochemiluminescence technology on patterned arrays for high sensitivity and a wide dynamic range The kit includes a 96-well plate detection antibodies wash buffers calibrators controls and other reagents needed to run the assay It can help diagnose recent or prior infection for the three viruses This multiplex viral antibody profiling kit enables efficient high-throughput testing from a single sample
Catalog No. 50-000-78267 Supplier Meso Scale Discovery Supplier No. K15347G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Viral Panel 3 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A and Influenza B in human serum and plasma This assay uses electrochemiluminescence technology on patterned arrays for high sensitivity and a wide dynamic range The kit includes a 96-well plate detection antibodies wash buffers calibrators controls and other reagents needed to run the assay It can help diagnose recent or prior infection for the three viruses This multiplex viral antibody profiling kit enables efficient high-throughput testing from a single sample