Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 3HMNKit
SDP
Catalog No. 5000082215
Encompass
The Meso Scale Discovery V-PLEX Plus Viral Panel 3 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A and Influenza B in human serum and plasma This assay utilizes electrochemiluminescence detection technology and provides quantitative results in under 5 hours The kit includes a 96-well plate coated with antigens for the viruses detection antibodies wash buffers controls calibrators and other necessary reagents for the assay This product can aid in the diagnosis of viral respiratory infections
Catalog No. 50-000-82215 Supplier Meso Scale Discovery Supplier No. K15347G2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Viral Panel 3 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A and Influenza B in human serum and plasma This assay utilizes electrochemiluminescence detection technology and provides quantitative results in under 5 hours The kit includes a 96-well plate coated with antigens for the viruses detection antibodies wash buffers controls calibrators and other necessary reagents for the assay This product can aid in the diagnosis of viral respiratory infections