Learn More
Meso Scale Discovery VPLEX Plus Viral PNL 3HMNKit
SDP
Catalog No. 5000079394
Encompass
The Meso Scale Discovery V-PLEX Plus Viral Panel 3 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A and Influenza B in human serum and plasma This assay kit includes a 96-well plate coated with antigens for the viruses detection antibodies standards controls diluent wash buffer read buffer and plate seals The electrochemiluminescent technology allows sensitive and accurate quantification of antibody levels This kit can help track immune responses to viral infections and vaccines With a testing time of just 3 5 hours this kit enables high-throughput COVID-19 and influenza serology testing
Catalog No. 50-000-79394 Supplier Meso Scale Discovery Supplier No. K15347G1
The Meso Scale Discovery V-PLEX Plus Viral Panel 3 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A and Influenza B in human serum and plasma This assay kit includes a 96-well plate coated with antigens for the viruses detection antibodies standards controls diluent wash buffer read buffer and plate seals The electrochemiluminescent technology allows sensitive and accurate quantification of antibody levels This kit can help track immune responses to viral infections and vaccines With a testing time of just 3 5 hours this kit enables high-throughput COVID-19 and influenza serology testing