Learn More
Meso Scale Discovery VPLEX Proinflam PNL 2 rat
SDP
Catalog No. 5000081696
Encompass
The Meso Scale Discovery V-PLEX Proinflammatory Panel 2 Rat Kit is used to simultaneously quantify 10 rat proinflammatory biomarkers This electrochemiluminescence (ECL) based assay allows researchers to measure IFN- IL-1 IL-2 IL-4 IL-5 IL-6 CXCL1 TNF- IL-13 and IL-10 from cell culture supernatants serum plasma (citrate heparin EDTA) and other biological samples The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Diluent and lyophilized SULFO-TAG labeled detection antibodies This multiplex immunoassay provides high sensitivity requires low sample volume and has a short 3 5 hour protocol
Catalog No. 50-000-81696 Supplier Meso Scale Discovery Supplier No. K15059D4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Proinflammatory Panel 2 Rat Kit is used to simultaneously quantify 10 rat proinflammatory biomarkers This electrochemiluminescence (ECL) based assay allows researchers to measure IFN- IL-1 IL-2 IL-4 IL-5 IL-6 CXCL1 TNF- IL-13 and IL-10 from cell culture supernatants serum plasma (citrate heparin EDTA) and other biological samples The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Diluent and lyophilized SULFO-TAG labeled detection antibodies This multiplex immunoassay provides high sensitivity requires low sample volume and has a short 3 5 hour protocol