Learn More
Meso Scale Discovery VPLEX Respiratory PNL 1 IgG
SDP
Catalog No. 5000079800
Encompass
The Meso Scale Discovery COVID-19 Respiratory Panel 1 IgG Assay Kit is an immunoassay that detects IgG antibodies to SARS-CoV-2 in human serum and plasma This qualitative assay uses MSD s electrochemiluminescence detection technology and provides results in under 5 hours The kit includes a 96-well plate coated with recombinant SARS-CoV-2 antigen detection antibody wash buffer read buffer calibrators and controls This assay aids in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-79800 Supplier Meso Scale Discovery Supplier No. K15365U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery COVID-19 Respiratory Panel 1 IgG Assay Kit is an immunoassay that detects IgG antibodies to SARS-CoV-2 in human serum and plasma This qualitative assay uses MSD s electrochemiluminescence detection technology and provides results in under 5 hours The kit includes a 96-well plate coated with recombinant SARS-CoV-2 antigen detection antibody wash buffer read buffer calibrators and controls This assay aids in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection