Learn More
Meso Scale Discovery VPLEX Respiratory PNL 1 IgM
SDP
Catalog No. 5000079802
Encompass
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a multiplex immunoassay that measures human interleukin 6 (IL-6) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators controls 10X diluent read buffer wash buffer and plate seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-79802 Supplier Meso Scale Discovery Supplier No. K15366U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a multiplex immunoassay that measures human interleukin 6 (IL-6) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators controls 10X diluent read buffer wash buffer and plate seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL