Learn More
Meso Scale Discovery VPLEX Respiratory PNL IgA
SDP
Catalog No. 5000079803
Encompass
The Meso Scale Diagnostics COVID-19 Respiratory Panel 1 IgA is an in vitro diagnostic test kit for the qualitative detection of IgA antibodies to SARS-CoV-2 in human serum and plasma This singleplex immunoassay uses electrochemiluminescence technology on Meso Scale Diagnostics proprietary MULTI-ARRAY platform It is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The test has 98 8% positive percent agreement and 99 6% negative percent agreement It is not intended to diagnose acute SARS-CoV-2 infection This product requires the MESO SECTOR S 600 instrument
Catalog No. 50-000-79803 Supplier Meso Scale Discovery Supplier No. K15367U2
The Meso Scale Diagnostics COVID-19 Respiratory Panel 1 IgA is an in vitro diagnostic test kit for the qualitative detection of IgA antibodies to SARS-CoV-2 in human serum and plasma This singleplex immunoassay uses electrochemiluminescence technology on Meso Scale Diagnostics proprietary MULTI-ARRAY platform It is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The test has 98 8% positive percent agreement and 99 6% negative percent agreement It is not intended to diagnose acute SARS-CoV-2 infection This product requires the MESO SECTOR S 600 instrument