Learn More
Meso Scale Discovery VPLEX Respiratory PNL IgG
SDP
Catalog No. 5000080348
Encompass
The Meso Scale Discovery COVID-19 Respiratory Panel 1 IgG Assay Kit is an electrochemiluminescent immunoassay that detects IgG antibodies to SARS-CoV-2 in human serum and plasma This qualitative assay can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes a 96-well plate coated with SARS-CoV-2 antigen detection antibody read buffer sample diluent calibrators and controls Results are analyzed on the MSD discovery workbench after a 2-hour incubation This assay has high sensitivity and specificity for SARS-CoV-2 IgG antibodies
Catalog No. 50-000-80348 Supplier Meso Scale Discovery Supplier No. K15365U2
The Meso Scale Discovery COVID-19 Respiratory Panel 1 IgG Assay Kit is an electrochemiluminescent immunoassay that detects IgG antibodies to SARS-CoV-2 in human serum and plasma This qualitative assay can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes a 96-well plate coated with SARS-CoV-2 antigen detection antibody read buffer sample diluent calibrators and controls Results are analyzed on the MSD discovery workbench after a 2-hour incubation This assay has high sensitivity and specificity for SARS-CoV-2 IgG antibodies