Learn More
Meso Scale Discovery VPLEX Respiratory PNL IgM
SDP
Catalog No. 5000079801
Encompass
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a highly sensitive and quantitative immunoassay for the detection of human interleukin 6 (IL-6) in cell culture supernatants serum plasma and other biological fluids The kit utilizes electrochemiluminescence technology and provides accurate reproducible results with a lower limit of detection of 0 05 pg/mL and an upper limit of 10 000 pg/mL The assay requires 50 L or less sample volume and can be read on the MSD QuickPlex SQ 120 MESO SECTOR S 600 or MESO QuickPlex SQ 120 instruments The kit includes a plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Diluent and lyophilized calibrator
Catalog No. 50-000-79801 Supplier Meso Scale Discovery Supplier No. K15366U2
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a highly sensitive and quantitative immunoassay for the detection of human interleukin 6 (IL-6) in cell culture supernatants serum plasma and other biological fluids The kit utilizes electrochemiluminescence technology and provides accurate reproducible results with a lower limit of detection of 0 05 pg/mL and an upper limit of 10 000 pg/mL The assay requires 50 L or less sample volume and can be read on the MSD QuickPlex SQ 120 MESO SECTOR S 600 or MESO QuickPlex SQ 120 instruments The kit includes a plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Diluent and lyophilized calibrator