Learn More
Meso Scale Discovery VPLEX SARSCoV2 384 PNL 1 IgA
SDP
Catalog No. 5000080003
Encompass
The V-PLEX Human IL-6 Kit is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of human interleukin 6 (IL-6) in serum plasma and cell culture supernatants The assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators diluents wash buffers read buffer and plate seals The assay has a typical sensitivity of 0 10 pg/mL and an upper limit of quantification of 1500 pg/mL for serum/plasma samples
Catalog No. 50-000-80003 Supplier Meso Scale Discovery Supplier No. K25394U4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human IL-6 Kit is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of human interleukin 6 (IL-6) in serum plasma and cell culture supernatants The assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators diluents wash buffers read buffer and plate seals The assay has a typical sensitivity of 0 10 pg/mL and an upper limit of quantification of 1500 pg/mL for serum/plasma samples