Learn More
Meso Scale Discovery VPLEX SARSCoV2 384 PNL 1 IgM
SDP
Catalog No. 5000081213
Encompass
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a multiplex immunoassay that measures levels of human interleukin-6 (IL-6) in serum plasma and cell culture supernatants This assay utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators controls 10X diluent read buffer wash buffer and plate seals This assay has high specificity and shows minimal cross-reactivity The kit has a detection range of 0 06-1500 pg/mL and requires 25 L of sample per well
Catalog No. 50-000-81213 Supplier Meso Scale Discovery Supplier No. K25393U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a multiplex immunoassay that measures levels of human interleukin-6 (IL-6) in serum plasma and cell culture supernatants This assay utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators controls 10X diluent read buffer wash buffer and plate seals This assay has high specificity and shows minimal cross-reactivity The kit has a detection range of 0 06-1500 pg/mL and requires 25 L of sample per well