Learn More
Meso Scale Discovery VPLEX SARSCoV2 384 PNL IgM
SDP
Catalog No. 5000079984
Encompass
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a highly sensitive and quantitative immunoassay for the detection of human interleukin 6 (IL-6) in cell culture supernatants serum plasma and other biological fluids This kit utilizes electrochemiluminescence technology and provides accurate reproducible results in just 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent read buffer wash buffer and plate seals The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-2500 pg/mL This kit allows for the rapid reliable quantification of IL-6 levels
Catalog No. 50-000-79984 Supplier Meso Scale Discovery Supplier No. K25393U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a highly sensitive and quantitative immunoassay for the detection of human interleukin 6 (IL-6) in cell culture supernatants serum plasma and other biological fluids This kit utilizes electrochemiluminescence technology and provides accurate reproducible results in just 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent read buffer wash buffer and plate seals The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-2500 pg/mL This kit allows for the rapid reliable quantification of IL-6 levels