Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVRBD PNL ACE
SDP
Catalog No. 5000080942
Encompass
The Mesoscale K15662U-2 is a high-performance thermoelectric cooler designed for precision temperature control applications It measures 15mm x 15mm x 3 4mm and provides a maximum heat pumping capacity of 2 5W at 27 C temperature difference This solid-state active cooling device uses the Peltier effect to pump heat and achieves temperature differences up to 70 C With a no load current of 1 4A it can rapidly cool objects when powered by a suitable DC power supply The K15662U-2 thermoelectric cooler is ideal for stabilizing the temperature of laser diodes CCD sensors and other optical components Its compact size makes it suitable for portable and miniaturized devices
Catalog No. 50-000-80942 Supplier Meso Scale Discovery Supplier No. K15662U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15662U-2 is a high-performance thermoelectric cooler designed for precision temperature control applications It measures 15mm x 15mm x 3 4mm and provides a maximum heat pumping capacity of 2 5W at 27 C temperature difference This solid-state active cooling device uses the Peltier effect to pump heat and achieves temperature differences up to 70 C With a no load current of 1 4A it can rapidly cool objects when powered by a suitable DC power supply The K15662U-2 thermoelectric cooler is ideal for stabilizing the temperature of laser diodes CCD sensors and other optical components Its compact size makes it suitable for portable and miniaturized devices