Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVRBD PNL ACE2
SDP
Catalog No. 5000080940
Encompass
The Meso Scale Discovery SARS-CoV-2 S1 RBD N501Y Kit is a research use only immunoassay that detects antibodies generated in response to the SARS-CoV-2 spike protein receptor binding domain containing the N501Y mutation This mutation is present in variants like B 1 1 7 B 1 351 and P 1 The kit contains a plate coated with recombinant SARS-CoV-2 S1 RBD N501Y antigen and all necessary reagents It can quantitatively measure IgG antibodies in serum plasma or other biological fluids in under 3 hours using an electrochemiluminescent detection method The kit has high sensitivity and specificity for assessing immune responses
Catalog No. 50-000-80940 Supplier Meso Scale Discovery Supplier No. K15662U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery SARS-CoV-2 S1 RBD N501Y Kit is a research use only immunoassay that detects antibodies generated in response to the SARS-CoV-2 spike protein receptor binding domain containing the N501Y mutation This mutation is present in variants like B 1 1 7 B 1 351 and P 1 The kit contains a plate coated with recombinant SARS-CoV-2 S1 RBD N501Y antigen and all necessary reagents It can quantitatively measure IgG antibodies in serum plasma or other biological fluids in under 3 hours using an electrochemiluminescent detection method The kit has high sensitivity and specificity for assessing immune responses