Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVRBD PNL IgA
SDP
Catalog No. 5000080966
Encompass
The Mesoscale K15661U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition module It has 4 analog voltage input channels with programmable gain amplifiers anti-aliasing filters and 16-bit ADCs The input voltage range is 10 V with software-selectable gains of 1 2 4 8 It has a USB interface and is compatible with Windows Linux and Mac operating systems The module is bus-powered for portability Key features include 250 kS/s sampling rate 16-bit resolution 4 analog input channels programmable gains anti-aliasing filters USB interface and cross-platform software
Catalog No. 50-000-80966 Supplier Meso Scale Discovery Supplier No. K15661U4
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15661U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition module It has 4 analog voltage input channels with programmable gain amplifiers anti-aliasing filters and 16-bit ADCs The input voltage range is 10 V with software-selectable gains of 1 2 4 8 It has a USB interface and is compatible with Windows Linux and Mac operating systems The module is bus-powered for portability Key features include 250 kS/s sampling rate 16-bit resolution 4 analog input channels programmable gains anti-aliasing filters USB interface and cross-platform software