Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVRBD PNL IgG
SDP
Catalog No. 5000078596
Encompass
The Meso Scale Discovery SARS-CoV-2 S1 RBD N501Y Protein is a recombinant protein corresponding to the receptor binding domain (RBD) of the SARS-CoV-2 spike protein with the N501Y mutation This protein can be used in immunoassays and as an immunogen The molecular weight is 25 8 kDa This product is supplied lyophilized from a 0 2 m filtered solution in PBS The recommended storage is at -20 C The protein is produced in HEK293 cells
Catalog No. 50-000-78596 Supplier Meso Scale Discovery Supplier No. K15659U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery SARS-CoV-2 S1 RBD N501Y Protein is a recombinant protein corresponding to the receptor binding domain (RBD) of the SARS-CoV-2 spike protein with the N501Y mutation This protein can be used in immunoassays and as an immunogen The molecular weight is 25 8 kDa This product is supplied lyophilized from a 0 2 m filtered solution in PBS The recommended storage is at -20 C The protein is produced in HEK293 cells