Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVRBD PNL IgG
SDP
Catalog No. 5000080835
Encompass
The Meso Scale Discovery SARS-CoV-2 S1 RBD N501Y Kit is a research use only immunoassay that detects antibodies generated in response to the SARS-CoV-2 spike protein receptor binding domain containing the N501Y mutation This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity wide dynamic range and rapid results The kit includes a 96-well plate coated with recombinant SARS-CoV-2 RBD protein detection antibody buffers calibrators and controls needed to run the assay It can help researchers monitor immune responses to SARS-CoV-2 variants containing the N501Y mutation
Catalog No. 50-000-80835 Supplier Meso Scale Discovery Supplier No. K15659U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery SARS-CoV-2 S1 RBD N501Y Kit is a research use only immunoassay that detects antibodies generated in response to the SARS-CoV-2 spike protein receptor binding domain containing the N501Y mutation This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity wide dynamic range and rapid results The kit includes a 96-well plate coated with recombinant SARS-CoV-2 RBD protein detection antibody buffers calibrators and controls needed to run the assay It can help researchers monitor immune responses to SARS-CoV-2 variants containing the N501Y mutation