Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVRBD PNL MsIgA
SDP
Catalog No. 5000078598
Encompass
The Mesoscale K15665U-2 is a high-performance thermoelectric module designed for cooling applications It uses bismuth telluride semiconductor material and can achieve a maximum temperature differential of 70 C This Peltier cooler has a maximum current of 15 7 Amps and a nominal voltage of 15 4 Volts With dimensions of 40 x 40 x 3 4mm this small module can provide up to 200 Watts of cooling power The K15665U-2 requires a heat sink and fan for proper thermal management Its ceramic substrates allow for high operating temperatures This thermoelectric cooler is well-suited for applications such as laser diode cooling
Catalog No. 50-000-78598 Supplier Meso Scale Discovery Supplier No. K15665U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15665U-2 is a high-performance thermoelectric module designed for cooling applications It uses bismuth telluride semiconductor material and can achieve a maximum temperature differential of 70 C This Peltier cooler has a maximum current of 15 7 Amps and a nominal voltage of 15 4 Volts With dimensions of 40 x 40 x 3 4mm this small module can provide up to 200 Watts of cooling power The K15665U-2 requires a heat sink and fan for proper thermal management Its ceramic substrates allow for high operating temperatures This thermoelectric cooler is well-suited for applications such as laser diode cooling