Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVRBD PNL MsIgG
SDP
Catalog No. 5000080967
Encompass
The Mesoscale K15663U-2 is a high-performance thermoelectric module that uses semiconductor thermocouples to convert heat flux directly into electrical power It operates as a solid-state heat pump or power generator and can be used for temperature control cooling heating and power generation applications This model has 127 thermocouples can handle up to 15W of heat pumping capacity and has an operating temperature range of -20 C to 71 C Some key specs are maximum T of 67 C voltage of 3 4V at Tmax and dimensions of 40 x 40 x 3 4mm The K15663U-2 requires a heat sink and cold side for operation
Catalog No. 50-000-80967 Supplier Meso Scale Discovery Supplier No. K15663U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15663U-2 is a high-performance thermoelectric module that uses semiconductor thermocouples to convert heat flux directly into electrical power It operates as a solid-state heat pump or power generator and can be used for temperature control cooling heating and power generation applications This model has 127 thermocouples can handle up to 15W of heat pumping capacity and has an operating temperature range of -20 C to 71 C Some key specs are maximum T of 67 C voltage of 3 4V at Tmax and dimensions of 40 x 40 x 3 4mm The K15663U-2 requires a heat sink and cold side for operation