Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVRBD PNL MsIgM
SDP
Catalog No. 5000080790
Encompass
The Meso Scale Discovery K15664U-2 is a 96-well plate kit for measuring human IL-6 in serum plasma cell culture supernatants and other biological samples It uses electrochemiluminescence (ECL) detection technology for high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators and buffers for sample dilution It has a detection range of 3 2-10 000 pg/mL and requires 25 L of sample per well The assay takes approximately 3 5 hours to complete It is designed for quantitative measurement of IL-6 levels using the MSD Sector Imager
Catalog No. 50-000-80790 Supplier Meso Scale Discovery Supplier No. K15664U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery K15664U-2 is a 96-well plate kit for measuring human IL-6 in serum plasma cell culture supernatants and other biological samples It uses electrochemiluminescence (ECL) detection technology for high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators and buffers for sample dilution It has a detection range of 3 2-10 000 pg/mL and requires 25 L of sample per well The assay takes approximately 3 5 hours to complete It is designed for quantitative measurement of IL-6 levels using the MSD Sector Imager