Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVSpik PNL ACE2
SDP
Catalog No. 5000080971
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Key Variant Spike Panel 1 (ACE2) Kit is a multiplex immunoassay measuring antibodies to the RBD of the SARS-CoV-2 spike protein including Alpha Beta Gamma Delta and Omicron variants It uses MSD electrochemiluminescence for high sensitivity a 6-log dynamic range and low sample volume Intended for qualitative detection of antibodies binding to RBD variants it aids in monitoring immune responses to COVID-19 vaccines and infection The kit includes a 96-well plate detection antibodies buffers calibrators controls and necessary accessories
Catalog No. 50-000-80971 Supplier Meso Scale Discovery Supplier No. K15654U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Key Variant Spike Panel 1 (ACE2) Kit is a multiplex immunoassay measuring antibodies to the RBD of the SARS-CoV-2 spike protein including Alpha Beta Gamma Delta and Omicron variants It uses MSD electrochemiluminescence for high sensitivity a 6-log dynamic range and low sample volume Intended for qualitative detection of antibodies binding to RBD variants it aids in monitoring immune responses to COVID-19 vaccines and infection The kit includes a 96-well plate detection antibodies buffers calibrators controls and necessary accessories