Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVSpike PNL
SDP
Catalog No. 5000079284
Encompass
The Meso Scale Diagnostics V-PLEX SARS-CoV-2 Key Variant Spike Panel 1 MSIGM Kit is an immunoassay that detects antibodies to the spike protein of SARS-CoV-2 variants in human serum plasma and dried blood spot samples This 25-plate kit includes reagents to run the assay on the MSD platform allowing for quantitative measurement of IgG antibodies binding to the spike proteins of the original Wuhan strain as well as the Alpha Beta Gamma Delta and Omicron variants The kit enables investigation of the antibody response to emerging SARS-CoV-2 variants
Catalog No. 50-000-79284 Supplier Meso Scale Discovery Supplier No. K15656U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Diagnostics V-PLEX SARS-CoV-2 Key Variant Spike Panel 1 MSIGM Kit is an immunoassay that detects antibodies to the spike protein of SARS-CoV-2 variants in human serum plasma and dried blood spot samples This 25-plate kit includes reagents to run the assay on the MSD platform allowing for quantitative measurement of IgG antibodies binding to the spike proteins of the original Wuhan strain as well as the Alpha Beta Gamma Delta and Omicron variants The kit enables investigation of the antibody response to emerging SARS-CoV-2 variants