Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVSpike PNL
SDP
Catalog No. 5000079303
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Key Variant Spike Panel 1 Mouse IgG Kit measures IgG antibodies to the RBD of the SARS-CoV-2 spike protein and its variants (Alpha Beta Gamma Delta) in mouse serum and plasma This electrochemiluminescence immunoassay detects IgG antibodies binding to the RBD offering insights into immune responses The kit includes a 96-well plate with RBD antigens detection antibodies calibrators controls and buffers Results are available in 3 5 hours using an MSD instrument aiding in monitoring antibody responses to vaccines or infections in mouse models
Catalog No. 50-000-79303 Supplier Meso Scale Discovery Supplier No. K15655U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Key Variant Spike Panel 1 Mouse IgG Kit measures IgG antibodies to the RBD of the SARS-CoV-2 spike protein and its variants (Alpha Beta Gamma Delta) in mouse serum and plasma This electrochemiluminescence immunoassay detects IgG antibodies binding to the RBD offering insights into immune responses The kit includes a 96-well plate with RBD antigens detection antibodies calibrators controls and buffers Results are available in 3 5 hours using an MSD instrument aiding in monitoring antibody responses to vaccines or infections in mouse models