Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVSpike PNL
SDP
Catalog No. 5000080864
Encompass
The Mesoscale K15657U-2 is a high-performance thermoelectric module that can be used for precision temperature control It contains 127 thermocouples and provides 15 4 Watts of cooling capacity at a hot side temperature of 27 C and a cold side temperature of 0 C The module operates on 15 Volts DC and can handle up to 8 Amps of current With dimensions of 40 x 40 x 3 4 mm the K15657U-2 module weighs approximately 85 grams This thermoelectric cooler is designed for regulated closed-loop temperature control applications
Catalog No. 50-000-80864 Supplier Meso Scale Discovery Supplier No. K15657U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15657U-2 is a high-performance thermoelectric module that can be used for precision temperature control It contains 127 thermocouples and provides 15 4 Watts of cooling capacity at a hot side temperature of 27 C and a cold side temperature of 0 C The module operates on 15 Volts DC and can handle up to 8 Amps of current With dimensions of 40 x 40 x 3 4 mm the K15657U-2 module weighs approximately 85 grams This thermoelectric cooler is designed for regulated closed-loop temperature control applications