Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVSpike PNL
SDP
Catalog No. 5000080929
Encompass
The V-PLEX SARS-CoV-2 Key Variant Spike Panel 1 Mouse IgG Kit from Meso Scale Diagnostics is a multiplex immunoassay for detecting and quantifying IgG antibodies against the SARS-CoV-2 spike protein and its variants including Alpha Beta Gamma Delta and Omicron in mouse serum and plasma It includes a 96-well plate with spike protein antigens detection antibodies calibrators controls diluent wash buffer read buffer and plate seals This electrochemiluminescence immunoassay provides sensitive and accurate IgG measurement aiding in evaluating immune responses to SARS-CoV-2 vaccines and infections
Catalog No. 50-000-80929 Supplier Meso Scale Discovery Supplier No. K15655U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Key Variant Spike Panel 1 Mouse IgG Kit from Meso Scale Diagnostics is a multiplex immunoassay for detecting and quantifying IgG antibodies against the SARS-CoV-2 spike protein and its variants including Alpha Beta Gamma Delta and Omicron in mouse serum and plasma It includes a 96-well plate with spike protein antigens detection antibodies calibrators controls diluent wash buffer read buffer and plate seals This electrochemiluminescence immunoassay provides sensitive and accurate IgG measurement aiding in evaluating immune responses to SARS-CoV-2 vaccines and infections