Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVSpike PNL
SDP
Catalog No. 5000081022
Encompass
The Meso Scale Discovery SARS-CoV-2 Spike B 1 1 7 V-PLEX Plus Kit is an electrochemiluminescence immunoassay that detects antibodies to the SARS-CoV-2 Spike protein containing the B 1 1 7 mutations This assay can help determine if someone has been previously infected specifically with the B 1 1 7 variant of SARS-CoV-2 The kit includes a 96-well plate coated with recombinant SARS-CoV-2 Spike protein detection antibodies wash buffer read buffer calibrators and controls Results are obtained in under 3 hours using Meso Scale Discovery instrumentation This product enables sensitive quantitative antibody detection
Catalog No. 50-000-81022 Supplier Meso Scale Discovery Supplier No. K15657U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery SARS-CoV-2 Spike B 1 1 7 V-PLEX Plus Kit is an electrochemiluminescence immunoassay that detects antibodies to the SARS-CoV-2 Spike protein containing the B 1 1 7 mutations This assay can help determine if someone has been previously infected specifically with the B 1 1 7 variant of SARS-CoV-2 The kit includes a 96-well plate coated with recombinant SARS-CoV-2 Spike protein detection antibodies wash buffer read buffer calibrators and controls Results are obtained in under 3 hours using Meso Scale Discovery instrumentation This product enables sensitive quantitative antibody detection