Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVSpike PNL
SDP
Catalog No. 5000081086
Encompass
The Mesoscale K15656U-2 is a high-performance thermoelectric cooler with a maximum heat pumping capacity of 15W It contains an array of 127 thermocouples and can achieve a maximum temperature differential of 70 C Some key features include its small footprint of 40 x 40 mm ability to precisely control temperature with 0 01 C resolution and integrated thermistor for temperature sensing With a high coefficient of performance and precise temperature control this Peltier cooler is well-suited for applications like analytical instrumentation industrial lasers photonics packaging and stabilizing the temperature of electronic components or small chambers
Catalog No. 50-000-81086 Supplier Meso Scale Discovery Supplier No. K15656U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15656U-2 is a high-performance thermoelectric cooler with a maximum heat pumping capacity of 15W It contains an array of 127 thermocouples and can achieve a maximum temperature differential of 70 C Some key features include its small footprint of 40 x 40 mm ability to precisely control temperature with 0 01 C resolution and integrated thermistor for temperature sensing With a high coefficient of performance and precise temperature control this Peltier cooler is well-suited for applications like analytical instrumentation industrial lasers photonics packaging and stabilizing the temperature of electronic components or small chambers