Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVSpike PNL ACE
SDP
Catalog No. 5000080970
Encompass
The V-PLEX SARS-CoV-2 Key Variant Spike Panel 1 (ACE2) Kit from Meso Scale Diagnostics is a multiplex immunoassay panel that allows for the simultaneous detection and quantitation of antibodies to the receptor binding domain (RBD) of the SARS-CoV-2 spike protein and its key variants (Alpha Beta Gamma Delta Omicron BA 1 Omicron BA 2/BA 5) The kit contains a 96-well plate coated with recombinant RBD antigens detection antibodies assay diluent wash buffer read buffer and calibrators and controls The electrochemiluminescence immunoassay allows sensitive and accurate measurement of IgG antibodies that block the interaction between RBD and ACE2 receptor
Catalog No. 50-000-80970 Supplier Meso Scale Discovery Supplier No. K15654U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Key Variant Spike Panel 1 (ACE2) Kit from Meso Scale Diagnostics is a multiplex immunoassay panel that allows for the simultaneous detection and quantitation of antibodies to the receptor binding domain (RBD) of the SARS-CoV-2 spike protein and its key variants (Alpha Beta Gamma Delta Omicron BA 1 Omicron BA 2/BA 5) The kit contains a 96-well plate coated with recombinant RBD antigens detection antibodies assay diluent wash buffer read buffer and calibrators and controls The electrochemiluminescence immunoassay allows sensitive and accurate measurement of IgG antibodies that block the interaction between RBD and ACE2 receptor