Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVSpike PNL IgA
SDP
Catalog No. 5000081016
Encompass
The Meso Scale Discovery K15653U-4 is a 96-well plate with a carbon electrode sensor for label-free detection of proteins via electrochemiluminescence (ECL) It offers high-sensitivity quantitative analysis and screening with low background and high signal-to-noise ratios The product includes 4 plates each allowing 96 assays Compatible with Meso Scale Discovery instruments and reagents it supports ECL-based immunoassays biomolecular interaction analysis and cell signaling studies featuring high sensitivity broad dynamic range and rapid assay times
Catalog No. 50-000-81016 Supplier Meso Scale Discovery Supplier No. K15653U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery K15653U-4 is a 96-well plate with a carbon electrode sensor for label-free detection of proteins via electrochemiluminescence (ECL) It offers high-sensitivity quantitative analysis and screening with low background and high signal-to-noise ratios The product includes 4 plates each allowing 96 assays Compatible with Meso Scale Discovery instruments and reagents it supports ECL-based immunoassays biomolecular interaction analysis and cell signaling studies featuring high sensitivity broad dynamic range and rapid assay times