Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVSpike PNL IgG
SDP
Catalog No. 5000080960
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Key Variant Spike Panel 1 IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to the spike protein of SARS-CoV-2 and its key variants in human serum and plasma This qualitative assay can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The panel allows for specific detection of antibodies to the spike proteins of the original Wuhan strain as well as the Alpha Beta Gamma Delta and Omicron variants Results are available in under 3 hours
Catalog No. 50-000-80960 Supplier Meso Scale Discovery Supplier No. K15651U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Key Variant Spike Panel 1 IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to the spike protein of SARS-CoV-2 and its key variants in human serum and plasma This qualitative assay can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The panel allows for specific detection of antibodies to the spike proteins of the original Wuhan strain as well as the Alpha Beta Gamma Delta and Omicron variants Results are available in under 3 hours