Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVSpike PNL IgM
SDP
Catalog No. 5000080833
Encompass
The Meso Scale Discovery SARS-CoV-2 Spike BA 2 Antigen Kit is a multiplex immunoassay that detects the SARS-CoV-2 Spike protein from the BA 2 Omicron variant in human serum plasma and whole blood samples This assay uses electrochemiluminescence technology and provides quantitative results in under 3 hours The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrators controls assay and wash buffers and plate seals This product allows for high-throughput testing of SARS-CoV-2 BA 2 variant samples
Catalog No. 50-000-80833 Supplier Meso Scale Discovery Supplier No. K15652U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery SARS-CoV-2 Spike BA 2 Antigen Kit is a multiplex immunoassay that detects the SARS-CoV-2 Spike protein from the BA 2 Omicron variant in human serum plasma and whole blood samples This assay uses electrochemiluminescence technology and provides quantitative results in under 3 hours The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrators controls assay and wash buffers and plate seals This product allows for high-throughput testing of SARS-CoV-2 BA 2 variant samples