Learn More
Meso Scale Discovery VPLEX SARSCoV2 KVSpike PNL IgM
SDP
Catalog No. 5000081051
Encompass
The Simoa SARS-CoV-2 Spike BA 2 Kit from Meso Scale Diagnostics is a highly sensitive immunoassay that quantitatively measures the SARS-CoV-2 Spike protein from the BA 2 Omicron variant in serum plasma whole blood dried blood spots and saliva This assay utilizes electrochemiluminescence detection and provides results in under 3 hours The kit includes a plate coated with antibodies that capture the Spike protein detection antibodies conjugated to an electrochemiluminescent label calibrators controls assay and sample diluents wash buffers and an MSD reader This assay can aid in understanding immune response to the BA 2 variant
Catalog No. 50-000-81051 Supplier Meso Scale Discovery Supplier No. K15652U2
Add to Cart
Add to Cart
Request a Quote
The Simoa SARS-CoV-2 Spike BA 2 Kit from Meso Scale Diagnostics is a highly sensitive immunoassay that quantitatively measures the SARS-CoV-2 Spike protein from the BA 2 Omicron variant in serum plasma whole blood dried blood spots and saliva This assay utilizes electrochemiluminescence detection and provides results in under 3 hours The kit includes a plate coated with antibodies that capture the Spike protein detection antibodies conjugated to an electrochemiluminescent label calibrators controls assay and sample diluents wash buffers and an MSD reader This assay can aid in understanding immune response to the BA 2 variant